Search
Search
| Gene Symbol: Cd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290802.1 | NP_001277731.1 | 3 - 4 | 306 | 60 | |
| NM_001290804.1 | NP_001277733.1 | 2 - 3 | 231 | 60 | ||
| NM_007645.4 | NP_031671.1 | 2 - 3 | 255 | 60 | ||
| GenBank mRNA | AK154250.1 | - | 2 - 3 | 292 | 60 | |
| AK154466.1 | - | 3 - 4 | 276 | 60 | ||
| AK155663.1 | - | 2 - 3 | 232 | 60 | ||
| AK155966.1 | - | 2 - 3 | 220 | 60 | ||
| BC019402.1 | - | 2 - 3 | 292 | 4013070 | 60 | |
| BE849269.1 | - | 2 - 3 | 173 | 60 | ||
| CT010217.1 | - | 2 - 3 | 144 | 60 | ||
| CX226822.1 | - | 3 - 4 | 307 | 60 | ||
Set Membership: |
|