Search
Search
| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011223.3 | NP_035353.1 | 7 - 8 | 1181 | 91 | |
| NM_133915.3 | NP_598676.2 | 8 - 9 | 1283 | 91 | ||
| GenBank mRNA | AF293882.1 | - | 7 - 8 | 1106 | 91 | |
| AF293883.1 | - | 7 - 8 | 1103 | 91 | ||
| AK148060.1 | - | 7 - 8 | 1167 | 91 | ||
| AK149933.1 | - | 7 - 8 | 1146 | 91 | ||
| AK167299.1 | - | 7 - 8 | 1120 | 91 | ||
| AK171226.1 | - | 7 - 8 | 1169 | 91 | ||
| AK171436.1 | - | 7 - 8 | 1146 | 91 | ||
| BC003298.1 | - | 3 - 4 | 464 | 91 | ||
| BC025493.1 | - | 7 - 8 | 1139 | 91 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK089534.1 | - | Chr. - - [-] on Build | 91 | |||
Set Membership: |
|