Search
Search
| Gene Symbol: Pnkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290764.1 | NP_001277693.1 | 2 - 3 | 315 | 61 | |
| NM_001290766.1 | NP_001277695.1 | 1 - 2 | 407 | 61 | ||
| NM_001290767.1 | NP_001277696.1 | 1 - 2 | 150 | 61 | ||
| NM_021549.3 | NP_067524.2 | 1 - 2 | 407 | 61 | ||
| GenBank mRNA | AF129451.1 | - | 1 - 2 | 149 | 61 | |
| AK049992.1 | - | 1 - 2 | 210 | 61 | ||
| AK159535.1 | - | 1 - 2 | 173 | 61 | ||
| AK159770.1 | - | 1 - 2 | 173 | 61 | ||
| AK160020.1 | - | 1 - 2 | 173 | 61 | ||
| AK160160.1 | - | 2 - 3 | 311 | 61 | ||
| AK172558.1 | - | 1 - 2 | 212 | 61 | ||
| BC047269.1 | - | 1 - 2 | 214 | 61 | ||
| BC057659.1 | - | 1 - 2 | 209 | 2865792 | 61 | |
| BE627794.1 | - | 1 - 2 | 372 | 61 | ||
| CK628081.1 | - | 1 - 2 | 219 | 61 | ||
| CX223963.1 | - | 1 - 2 | 394 | 61 | ||
Set Membership: |
|