Search
Search

Tspan14| Gene Symbol: Tspan14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145928.1 | NP_666040.1 | 7 - 8 | 673 | 118 | |
| GenBank mRNA | AK030593.1 | - | 7 - 8 | 675 | 118 | |
| AK156671.1 | - | 7 - 8 | 686 | 118 | ||
| BC024611.1 | - | 7 - 8 | 678 | 4188261 | 118 | |
| BC025568.1 | - | 7 - 8 | 687 | 5322531 | 118 | |
| BC026574.1 | - | 7 - 8 | 673 | 4954874 | 118 | |
Set Membership: |
|