Search
Search

Adam32| Gene Symbol: Adam32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293693.1 | NP_001280622.1 | 7 - 8 | 599 | 151 | |
| NM_153397.2 | NP_700446.2 | 7 - 8 | 599 | 151 | ||
| GenBank mRNA | AF513715.1 | - | 7 - 8 | 582 | 151 | |
| AK005759.1 | - | 7 - 8 | 590 | 151 | ||
| BC060983.1 | - | 7 - 8 | 596 | 6772485 | 151 | |
Set Membership: |
|