Search
Search

Gabrr2| Gene Symbol: Gabrr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008076.3 | NP_032102.2 | 7 - 8 | 1026 | 90 | |
| GenBank mRNA | AF024621.1 | - | 7 - 8 | 1139 | 90 | |
| AK132642.1 | - | 7 - 8 | 854 | 90 | ||
| AK160446.1 | - | 5 - 6 | 693 | 90 | ||
| BC057957.1 | - | 7 - 8 | 1011 | 5357713 | 90 | |
Set Membership: |
|