Search
Search
| Gene Symbol: Chat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009891.2 | NP_034021.1 | 11 - 12 | 1513 | 133 | |
| GenBank mRNA | AK046406.1 | - | 11 - 12 | 1514 | 133 | |
| AK047708.1 | - | 10 - 11 | 1405 | 133 | ||
| BC119322.1 | - | 10 - 11 | 1275 | 8734071 | 133 | |
| BC120589.1 | - | 10 - 11 | 1275 | 40129512 | 133 | |
Set Membership: |
|