Search
Search

Tmtc4| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028651.2 | NP_082927.1 | 11 - 12 | 1586 | 72 | |
| GenBank mRNA | AK017580.1 | - | 1 - 2 | 115 | 72 | |
| AK028333.1 | - | 11 - 12 | 1567 | 72 | ||
| AK033947.1 | - | 11 - 12 | 1587 | 72 | ||
| AK043293.1 | - | 11 - 12 | 1613 | 72 | ||
| AK044391.1 | - | 11 - 12 | 1542 | 72 | ||
| AK045271.1 | - | 11 - 12 | 1548 | 72 | ||
| AK049748.1 | - | 12 - 13 | 1727 | 72 | ||
| AK082526.1 | - | 10 - 11 | 1414 | 72 | ||
| BC031368.1 | - | 7 - 8 | 960 | 5029461 | 72 | |
Set Membership: |
|