Search
Search

Sema3a| Gene Symbol: Sema3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243072.1 | NP_001230001.1 | 14 - 15 | 1876 | 64 | |
| NM_001243073.1 | NP_001230002.1 | 14 - 15 | 1716 | 64 | ||
| NM_009152.4 | NP_033178.2 | 13 - 14 | 2144 | 64 | ||
| GenBank mRNA | AK140817.1 | - | 14 - 15 | 1717 | 64 | |
| BC057588.1 | - | 13 - 14 | 1791 | 6416990 | 64 | |
| BC090844.1 | - | 14 - 15 | 1876 | 30532393 | 64 | |
| D85028.2 | - | 13 - 14 | 2146 | 64 | ||
| L40484.1 | - | 10 - 11 | 1177 | 64 | ||
| L41541.1 | - | 13 - 14 | 1495 | 64 | ||
| X85993.1 | - | 13 - 14 | 1605 | 64 | ||
Set Membership: |
|