Search
Search

Slc9a4| Gene Symbol: Slc9a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177084.3 | NP_796058.1 | 6 - 7 | 1771 | 100 | |
| GenBank mRNA | AK085681.1 | - | 6 - 7 | 1797 | 100 | |
| AK141534.1 | - | 6 - 7 | 1772 | 100 | ||
| AK170522.1 | - | 4 - 5 | 610 | 100 | ||
| BC120543.1 | - | 6 - 7 | 1689 | 40129466 | 100 | |
Set Membership: |
|