Search
Search
| Gene Symbol: Stac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016853.2 | NP_058549.1 | 6 - 7 | 872 | 103 | |
| GenBank mRNA | AK134777.1 | - | 5 - 6 | 602 | 103 | |
| AK143440.1 | - | 6 - 7 | 905 | 103 | ||
| BC107325.1 | - | 6 - 7 | 796 | 40054255 | 103 | |
| BC107326.1 | - | 6 - 7 | 796 | 40054256 | 103 | |
| D86639.1 | - | 6 - 7 | 872 | 103 | ||
Set Membership: |
|