Search
Search

Psmf1| Gene Symbol: Psmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031363.1 | - | 2 - 3 | 439 | 57 | |
| AK045891.1 | - | 2 - 3 | 422 | 57 | ||
| AK053924.1 | - | 2 - 3 | 440 | 57 | ||
| AK078867.1 | - | 2 - 3 | 398 | 57 | ||
| BC012260.1 | - | 2 - 3 | 401 | 57 | ||
| BC128314.1 | - | 1 - 2 | 184 | 57 | ||
| BC131928.1 | - | 2 - 3 | 303 | 40130205 | 57 | |
| BC131930.1 | - | 2 - 3 | 303 | 40130207 | 57 | |
Set Membership: |
|