Search
Search

Smtnl2| Gene Symbol: Smtnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177776.3 | NP_808444.1 | 5 - 6 | 1058 | 84 | |
| GenBank mRNA | AK051585.1 | - | 5 - 6 | 1058 | 84 | |
| BC037993.1 | - | 5 - 6 | 1056 | 5064159 | 84 | |
Set Membership: |
|