Search
Search

Serpinf2| Gene Symbol: Serpinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008878.2 | NP_032904.1 | 2 - 3 | 112 | 65 | |
| GenBank mRNA | AK077563.1 | - | 2 - 3 | 115 | 65 | |
| AK090208.1 | - | 2 - 3 | 96 | 65 | ||
| BC026756.1 | - | 2 - 3 | 86 | 5124817 | 65 | |
| GQ896353.1 | - | 1 - 2 | 63 | 65 | ||
| GQ896354.1 | - | 1 - 2 | 63 | 65 | ||
| Z36774.1 | - | 2 - 3 | 99 | 65 | ||
Set Membership: |
|