Search
Search
| Gene Symbol: Car7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301164.1 | NP_001288093.1 | 1 - 2 | 520 | 67 | |
| GenBank mRNA | BC038270.1 | - | 1 - 2 | 495 | 67 | |
| BY741075.1 | - | 1 - 2 | 520 | 67 | ||
Set Membership: |
|