Search
Search

March10| Gene Symbol: March10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039242.2 | NP_001034331.1 | 2 - 3 | 343 | 146 | |
| NM_172568.2 | NP_766156.2 | 2 - 3 | 343 | 146 | ||
| GenBank mRNA | AK132734.1 | - | 2 - 3 | 332 | 146 | |
| BC100474.1 | - | 2 - 3 | 336 | 30918723 | 146 | |
Set Membership: |
|