Search
Search

Spire2| Gene Symbol: Spire2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172287.2 | NP_758491.1 | 3 - 4 | 705 | 93 | |
| GenBank mRNA | BC026502.1 | - | 2 - 3 | 533 | 4506974 | 93 |
| BC049152.1 | - | 3 - 4 | 705 | 6311485 | 93 | |
| BC054072.1 | - | 3 - 4 | 705 | 93 | ||
Set Membership: |
|