| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100039079.mm.1 |
NM_030206.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039079.mm.1 |
NM_030206.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039079.mm.1 |
TC1100004123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100039077.mm.1 |
NM_030206.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039077.mm.1 |
NM_030206.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039077.mm.1 |
TC1100004123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100039078.mm.1 |
NM_030206.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039078.mm.1 |
NM_030206.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039078.mm.1 |
TC1100004123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100039081.mm.1 |
NM_030206.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039081.mm.1 |
NM_030206.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039081.mm.1 |
TC1100004123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100039082.mm.1 |
NM_030206.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039082.mm.1 |
NM_030206.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039082.mm.1 |
TC1100004123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100039080.mm.1 |
NM_030206.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039080.mm.1 |
NM_030206.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039080.mm.1 |
TC1100004123.mm.1 |
MTA TC |