Search
Search
| Gene Symbol: Vil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009509.2 | NP_033535.2 | 15 - 16 | 1893 | 95 | |
| GenBank mRNA | AK027908.1 | - | 15 - 16 | 1890 | 95 | |
| BC015267.1 | - | 15 - 16 | 1869 | 4236751 | 95 | |
| BC117875.2 | - | 15 - 16 | 1858 | 40095718 | 95 | |
| BC145671.1 | - | 15 - 16 | 1860 | 40130974 | 95 | |
| M98454.1 | - | 15 - 16 | 1883 | 95 | ||
Set Membership: |
|