Search
Search
| Gene Symbol: Dab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010014.3 | NP_034144.1 | 4 - 5 | 617 | 137 | |
| NM_177259.4 | NP_796233.2 | 4 - 5 | 617 | 137 | ||
| NR_104385.1 | 4 - 5 | 617 | 137 | |||
| GenBank mRNA | AK005640.1 | - | 3 - 4 | 428 | 137 | |
| BC051993.1 | - | 6 - 7 | 750 | 137 | ||
| BC138538.1 | - | 3 - 4 | 338 | 8861558 | 137 | |
| BC138539.1 | - | 3 - 4 | 338 | 8861559 | 137 | |
| BY731247.1 | - | 4 - 5 | 584 | 137 | ||
| CO040161.1 | - | 4 - 5 | 137 | |||
| Y08379.1 | - | 4 - 5 | 565 | 137 | ||
| Y08380.1 | - | 4 - 5 | 565 | 137 | ||
| Y08381.1 | - | 4 - 5 | 565 | 137 | ||
Set Membership: |
|