Search
Search

Haus6| Gene Symbol: Haus6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173400.2 | NP_775576.2 | 6 - 7 | 858 | 91 | |
| GenBank mRNA | AK051016.1 | - | 6 - 7 | 879 | 91 | |
| AK169222.1 | - | 6 - 7 | 860 | 91 | ||
| AK173206.1 | - | 6 - 7 | 830 | 91 | ||
| BC064725.1 | - | 1 - 2 | 55 | 91 | ||
| BC068237.1 | - | 6 - 7 | 858 | 30550558 | 91 | |
Set Membership: |
|