Search
Search

Frmd3| Gene Symbol: Frmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163732.1 | NP_001157204.1 | 3 - 4 | 466 | 85 | |
| NM_172869.4 | NP_766457.1 | 3 - 4 | 466 | 85 | ||
| GenBank mRNA | AK034956.1 | - | 3 - 4 | 441 | 85 | |
| AK048561.1 | - | 3 - 4 | 409 | 85 | ||
| AK079349.1 | - | 3 - 4 | 467 | 85 | ||
| BC037727.1 | - | 3 - 4 | 637 | 85 | ||
| BC112419.1 | - | 3 - 4 | 374 | 40061426 | 85 | |
| BC113795.1 | - | 3 - 4 | 374 | 40061428 | 85 | |
| BC118964.2 | - | 3 - 4 | 376 | 40097059 | 85 | |
Set Membership: |
|