Search
Search
| Gene Symbol: Ipo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181517.3 | NP_852658.2 | 6 - 7 | 719 | 124 | |
| GenBank mRNA | AK039534.1 | - | 6 - 7 | 871 | 124 | |
| AK148820.1 | - | 6 - 7 | 830 | 124 | ||
| AY316153.1 | - | 6 - 7 | 719 | 124 | ||
| BC053524.1 | - | 3 - 4 | 402 | 124 | ||
Set Membership: |
|