Search
Search

Mep1a| Gene Symbol: Mep1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008585.2 | NP_032611.2 | 8 - 9 | 831 | 84 | |
| GenBank mRNA | AK143906.1 | - | 8 - 9 | 789 | 84 | |
| AK164734.1 | - | 8 - 9 | 786 | 84 | ||
| BC015258.1 | - | 8 - 9 | 831 | 4241252 | 84 | |
| M74897.1 | - | 8 - 9 | 830 | 84 | ||
Set Membership: |
|