Search
Search

Mep1a| Gene Symbol: Mep1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008585.2 | NP_032611.2 | 9 - 10 | 984 | 62 | |
| GenBank mRNA | AK093914.1 | - | 9 - 10 | 943 | 62 | |
| AK143906.1 | - | 9 - 10 | 942 | 62 | ||
| AK164734.1 | - | 9 - 10 | 939 | 62 | ||
| BC015258.1 | - | 9 - 10 | 984 | 4241252 | 62 | |
| M74897.1 | - | 9 - 10 | 983 | 62 | ||
Set Membership: |
|