Search
Search
| Gene Symbol: Dlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286013.1 | NP_001272942.1 | 4 - 5 | 471 | 82 | |
| NM_001286028.1 | NP_001272957.1 | 4 - 5 | 358 | 82 | ||
| GenBank mRNA | BC118057.1 | - | 4 - 5 | 358 | 40105170 | 82 |
| BC122518.1 | - | 5 - 6 | 386 | 40105175 | 82 | |
| BG868693.1 | - | 4 - 5 | 451 | 82 | ||
| FM180474.1 | - | 4 - 5 | 471 | 82 | ||
Set Membership: |
|