Search
Search

Rabgap1l| Gene Symbol: Rabgap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013862.5 | NP_038890.3 | 18 - 19 | 2451 | 90 | |
| GenBank mRNA | AK035796.1 | - | 18 - 19 | 2452 | 90 | |
| BC145811.1 | - | 18 - 19 | 2296 | 40131114 | 90 | |
| BC145813.1 | - | 18 - 19 | 2296 | 40131116 | 90 | |
Set Membership: |
|