Search
Search

Nova1| Gene Symbol: Nova1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021361.1 | NP_067336.1 | 3 - 4 | 466 | 75 | |
| GenBank mRNA | AF232828.1 | - | 3 - 4 | 400 | 75 | |
| AW491473.1 | - | 2 - 3 | 75 | |||
Set Membership: |
|