Search
Search

Xylt2| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145828.3 | NP_665827.2 | 5 - 6 | 1170 | 70 | |
| GenBank mRNA | AJ291751.2 | - | 5 - 6 | 1087 | 70 | |
| AK083482.1 | - | 5 - 6 | 1139 | 70 | ||
| AK086804.1 | - | 5 - 6 | 1102 | 70 | ||
| AK158056.1 | - | 5 - 6 | 1170 | 70 | ||
| AK177270.1 | - | 2 - 3 | 188 | 70 | ||
| BC034082.1 | - | 4 - 5 | 689 | 4239646 | 70 | |
Set Membership: |
|