Search
Search

Scn8a| Gene Symbol: Scn8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077499.2 | NP_001070967.1 | 2 - 3 | 427 | 84 | |
| NM_011323.3 | NP_035453.2 | 1 - 2 | 328 | 84 | ||
| GenBank mRNA | AF049617.1 | - | 1 - 2 | 274 | 84 | |
| AK158609.1 | - | 2 - 3 | 389 | 84 | ||
| U26707.1 | - | 1 - 2 | 286 | 84 | ||
Set Membership: |
|