Search
Search
| Gene Symbol: Relb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290457.1 | NP_001277386.1 | 9 - 10 | 1395 | 99 | |
| NM_009046.2 | NP_033072.2 | 10 - 11 | 1404 | 99 | ||
| GenBank mRNA | AK146932.1 | - | 10 - 11 | 1405 | 99 | |
| AK156767.1 | - | 10 - 11 | 1360 | 99 | ||
| BC019765.1 | - | 10 - 11 | 1404 | 3584200 | 99 | |
| BC034523.1 | - | 9 - 10 | 1208 | 4485540 | 99 | |
| BC117792.2 | - | 2 - 3 | 99 | |||
| BC117793.1 | - | 10 - 11 | 1340 | 40092162 | 99 | |
| DQ020178.1 | - | 9 - 10 | 1056 | 99 | ||
| DQ020179.1 | - | 9 - 10 | 2172 | 99 | ||
| DQ020181.1 | - | 10 - 11 | 2278 | 99 | ||
| M83380.1 | - | 10 - 11 | 1241 | 99 | ||
Set Membership: |
|