Search
Search
| Gene Symbol: Mpp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019579.3 | NP_062525.1 | 10 - 11 | 1702 | 156 | |
| GenBank mRNA | AF199008.1 | - | 10 - 11 | 1633 | 156 | |
| AK032731.1 | - | 10 - 11 | 1660 | 156 | ||
| AK133187.1 | - | 10 - 11 | 1595 | 156 | ||
| AK162014.1 | - | 10 - 11 | 1660 | 156 | ||
| BC067404.1 | - | 10 - 11 | 1563 | 156 | ||
| BC138624.1 | - | 10 - 11 | 1581 | 8861644 | 156 | |
Set Membership: |
|