Search
Search

Manba| Gene Symbol: Manba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027288.3 | NP_081564.3 | 7 - 8 | 1026 | 92 | |
| GenBank mRNA | AF306557.1 | - | 7 - 8 | 1030 | 92 | |
| AK036505.1 | - | 8 - 9 | 1126 | 92 | ||
| AK047057.1 | - | 7 - 8 | 1019 | 92 | ||
| AK142165.1 | - | 7 - 8 | 991 | 92 | ||
| AK154172.1 | - | 7 - 8 | 1008 | 92 | ||
| AK166022.1 | - | 7 - 8 | 1009 | 92 | ||
| AK166340.1 | - | 7 - 8 | 1024 | 92 | ||
| AK170290.1 | - | 7 - 8 | 1023 | 92 | ||
| BC031409.1 | - | 7 - 8 | 969 | 4952710 | 92 | |
Set Membership: |
|