Search
Search

Ncoa1| Gene Symbol: Ncoa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010881.2 | NP_035011.1 | 9 - 10 | 1520 | 82 | |
| GenBank mRNA | BC068177.1 | - | 6 - 7 | 828 | 30628581 | 82 |
| BC080866.1 | - | 9 - 10 | 1283 | 30532122 | 82 | |
| U56920.1 | - | 8 - 9 | 1078 | 82 | ||
| U64606.1 | - | 6 - 7 | 823 | 82 | ||
| U64828.1 | - | 9 - 10 | 1433 | 82 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK089302.1 | - | Chr. - - [-] on Build | 82 | |||
Set Membership: |
|