Search
Search

Atp1b4| Gene Symbol: Atp1b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290389.1 | NP_001277318.1 | 3 - 4 | 581 | 70 | |
| NM_133690.3 | NP_598451.1 | 3 - 4 | 593 | 70 | ||
| GenBank mRNA | AF348324.1 | - | 3 - 4 | 448 | 70 | |
| AF348325.1 | - | 3 - 4 | 436 | 70 | ||
| AK053500.1 | - | 3 - 4 | 532 | 70 | ||
| BC137923.1 | - | 3 - 4 | 534 | 8860942 | 70 | |
| BC137924.1 | - | 3 - 4 | 546 | 8860943 | 70 | |
Set Membership: |
|