Search
Search

Ambra1| Gene Symbol: Ambra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080754.1 | NP_001074223.1 | 15 - 16 | 3065 | 80 | |
| NM_172669.3 | NP_766257.3 | 14 - 15 | 3338 | 80 | ||
| GenBank mRNA | AK129433.1 | - | 10 - 11 | 2602 | 80 | |
| AK155513.1 | - | 14 - 15 | 3301 | 80 | ||
| AK169280.1 | - | 15 - 16 | 3018 | 80 | ||
| BC032293.1 | - | 15 - 16 | 2988 | 80 | ||
| BC054082.1 | - | 14 - 15 | 3271 | 80 | ||
Set Membership: |
|