Search
Search

Grb14| Gene Symbol: Grb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016719.1 | NP_057928.1 | 4 - 5 | 715 | 103 | |
| GenBank mRNA | AF155647.1 | - | 4 - 5 | 715 | 103 | |
| AK010849.1 | - | 4 - 5 | 732 | 103 | ||
| AK010903.1 | - | 4 - 5 | 732 | 103 | ||
| AK147041.1 | - | 4 - 5 | 724 | 103 | ||
| BC138581.1 | - | 4 - 5 | 621 | 8861601 | 103 | |
| BC145913.1 | - | 4 - 5 | 621 | 40131216 | 103 | |
Set Membership: |
|