Search
Search

Smek1| Gene Symbol: Smek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160214.1 | NP_001153686.1 | 4 - 5 | 1739 | 154 | |
| NM_211355.2 | NP_997594.1 | 4 - 5 | 1739 | 154 | ||
| GenBank mRNA | AK051239.1 | - | 4 - 5 | 1664 | 154 | |
| AK152480.1 | - | 4 - 5 | 1736 | 154 | ||
| AK156272.1 | - | 4 - 5 | 1701 | 154 | ||
| AK173331.1 | - | 4 - 5 | 1642 | 154 | ||
| BC064465.1 | - | 4 - 5 | 1437 | 6825400 | 154 | |
Set Membership: |
|