Search
Search
| Gene Symbol: Oog4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173773.1 | NP_776134.1 | 3 - 4 | 510 | 104 | |
| GenBank mRNA | AK054346.1 | - | 3 - 4 | 532 | 104 | |
| AY573564.1 | - | 3 - 4 | 510 | 104 | ||
| BC053722.1 | - | 2 - 3 | 381 | 104 | ||
| BC098485.1 | - | 2 - 3 | 369 | 30051767 | 104 | |
Set Membership: |
|