Search
Search

Crybg3| Gene Symbol: Crybg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174848.3 | NP_777273.2 | 8 - 9 | 2379 | 65 | |
| GenBank mRNA | BC043118.1 | - | 8 - 9 | 2379 | 6408859 | 65 |
Set Membership: |
|