Search
Search

Fbxo10| Gene Symbol: Fbxo10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024142.1 | NP_001019313.1 | 8 - 9 | 2305 | 62 | |
| GenBank mRNA | AK142389.1 | - | 7 - 8 | 2229 | 62 | |
| BC052485.1 | - | 7 - 8 | 1772 | 62 | ||
| BC054731.1 | - | 8 - 9 | 2295 | 62 | ||
| BC062126.1 | - | 7 - 8 | 1663 | 62 | ||
| BC079571.1 | - | 2 - 3 | 421 | 62 | ||
| BC094565.1 | - | 7 - 8 | 2089 | 6836944 | 62 | |
Set Membership: |
|