Search
Search

Rufy3| Gene Symbol: Rufy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289774.1 | NP_001276703.1 | 6 - 7 | 1010 | 119 | |
| NM_001289775.1 | NP_001276704.1 | 6 - 7 | 1010 | 119 | ||
| NM_001289776.1 | NP_001276705.1 | 7 - 8 | 1432 | 119 | ||
| NM_001289777.1 | NP_001276706.1 | 8 - 9 | 1257 | 119 | ||
| NM_027530.3 | NP_081806.1 | 6 - 7 | 1378 | 119 | ||
| GenBank mRNA | AK013185.1 | - | 8 - 9 | 1258 | 119 | |
| AK018184.1 | - | 6 - 7 | 1356 | 119 | ||
| AK083242.1 | - | 6 - 7 | 1313 | 119 | ||
| AK148795.1 | - | 6 - 7 | 674 | 119 | ||
| AK160820.1 | - | 6 - 7 | 1011 | 119 | ||
| BC049127.1 | - | 6 - 7 | 1317 | 6821567 | 119 | |
| BC058259.1 | - | 7 - 8 | 1151 | 6469137 | 119 | |
| CT010404.1 | - | 6 - 7 | 751 | 119 | ||
Set Membership: |
|