| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
NM_001252645.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
NM_001252645.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
NM_001252646.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
NM_001252646.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
NM_001252647.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
NM_001252647.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
NM_001252648.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
NM_001252648.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
NM_133797.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
NM_133797.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012688.mm.1 |
TC1300002034.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
NM_001252645.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
NM_001252645.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
NM_001252646.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
NM_001252646.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
NM_001252647.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
NM_001252647.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
NM_001252648.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
NM_001252648.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
NM_133797.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
NM_133797.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012696.mm.1 |
TC1300002034.mm.1 |
MTA TC |