| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325435 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325439 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325438 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325436 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325432 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325434 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325437 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325433 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2046184 |
BC034878.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2046184 |
AK084673.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2046184 |
AK009370.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2046184 |
NM_001162918.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2046184 |
NM_025515.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2144975 |
BC034878.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2144975 |
BB617939.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2144975 |
NM_001162918.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2144975 |
NM_025515.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2144975 |
AI643175.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
AK028362.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
AK044479.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
AK132746.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
AK150949.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
AK151555.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
AK168858.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
AK169106.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
BC026459.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
BC083132.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
NM_001025371.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757887 |
NM_011354.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463798 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
18216 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726677 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
710978 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
240598 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
288304 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
287691 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
588451 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
224134 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
172600 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
157519 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P118083 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P144061 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2144975 |
BC034878.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2144975 |
BB617939.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2144975 |
NM_001162918.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2144975 |
NM_025515.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2144975 |
AI643175.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2046184 |
BC034878.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2046184 |
AK084673.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2046184 |
AK009370.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2046184 |
NM_001162918.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2046184 |
NM_025515.3 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325432 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325420 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325418 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325423 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325429 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325439 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325443 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325426 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325417 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325437 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325431 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325446 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325452 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325448 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325435 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325441 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325425 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325451 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325444 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325440 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325438 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25325442 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P144061 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06281725 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06281743 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P665242 |
BC034878.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P665242 |
BC115973.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P665242 |
AK084673.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P665242 |
AK160230.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P665242 |
AK009370.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P131907 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P112437 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P130713 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P144061 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06588483 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06588469 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06588465 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06588460 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06588463 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06588481 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325439 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325437 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325435 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325436 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325434 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325433 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325432 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25325438 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325467 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325540 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325425 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325451 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325460 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325549 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325453 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325528 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325475 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325511 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325431 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325484 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325429 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325491 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325487 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325551 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325479 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325432 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325494 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325532 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325427 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325545 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325553 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325490 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325476 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325525 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325502 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325510 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325446 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325462 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325423 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325547 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325505 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325454 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325498 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325474 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325507 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325437 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325560 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325508 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325469 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325465 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325538 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325480 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325542 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325530 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325459 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325448 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325442 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325535 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325529 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325555 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325496 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325457 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325482 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325417 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325435 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325440 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325426 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325483 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325444 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325506 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325452 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325441 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325544 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325471 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325526 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325558 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25325534 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325484 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325444 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325438 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325446 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06028670 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325474 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325418 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325460 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325426 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325423 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325522 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325451 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325440 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325490 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325467 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325483 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325487 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325413 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325453 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325452 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06588469 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325459 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325435 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06655396 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325472 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325469 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325449 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325427 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325476 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325462 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06079478 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06655393 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325523 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325443 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325525 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06588478 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325448 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325479 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325417 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325524 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325454 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325414 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325412 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325482 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325441 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325465 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06588465 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325442 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325429 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06588460 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325457 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25325475 |
- |
- |