| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10410198 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10608663 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1437299_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10608663 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10410187 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10549949 |
- |
- |
| Mouse Genome 430 2.0 Array |
1437299_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1436794_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1436794_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1437299_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0900014107.mm.1 |
NM_001004145.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014107.mm.1 |
TC0900001678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900014105.mm.1 |
NM_001004145.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014105.mm.1 |
TC0900001678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900014104.mm.1 |
NM_001004145.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014104.mm.1 |
TC0900001678.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1437299_PM_at |
- |
- |
| Mouse Expression Set 430 |
1437299_at |
- |
- |
| Murine Genome U74v2 Set |
137632_f_at |
- |
- |
| Murine Genome U74v2 Set |
137631_f_at |
- |
- |
| Murine Genome U74v2 Set |
137631_PM_f_at |
- |
- |
| Murine Genome U74v2 Set |
137632_PM_f_at |
- |
- |