Search
Search

Ube3a| Gene Symbol: Ube3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033962.1 | NP_001029134.1 | 5 - 6 | 2247 | 157 | |
| NM_011668.2 | NP_035798.2 | 7 - 8 | 2434 | 157 | ||
| NM_173010.3 | NP_766598.1 | 5 - 6 | 2249 | 157 | ||
| GenBank mRNA | AK029133.1 | - | 5 - 6 | 2250 | 157 | |
| AK083795.1 | - | 5 - 6 | 2179 | 157 | ||
| AK220549.1 | - | 5 - 6 | 2147 | 157 | ||
| BC131658.1 | - | 5 - 6 | 2235 | 40110636 | 157 | |
| BC131659.1 | - | 5 - 6 | 2235 | 40110637 | 157 | |
Set Membership: |
|