Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008299.3 | NP_032325.2 | 1 | 547 | 99 | |
GenBank mRNA | AK005688.1 | - | 1 | 505 | 99 | |
BC048490.1 | - | 1 | 515 | 6775233 | 99 | |
U95607.1 | - | 1 | 549 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |