| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800020594.mm.1 |
NR_033641.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020594.mm.1 |
NR_110452.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020594.mm.1 |
TC0800002694.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020596.mm.1 |
NR_033641.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020596.mm.1 |
NR_110452.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020596.mm.1 |
TC0800002694.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020597.mm.1 |
NR_033641.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020597.mm.1 |
NR_110452.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020597.mm.1 |
TC0800002694.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020595.mm.1 |
NR_033641.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020595.mm.1 |
NR_110452.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020595.mm.1 |
TC0800002694.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020599.mm.1 |
NR_033641.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020599.mm.1 |
NR_110452.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020599.mm.1 |
TC0800002694.mm.1 |
MTA TC |