Search
Search

l7Rn6| Gene Symbol: l7Rn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_111916.1 | 1 | 2521 | 79 | ||
| GenBank mRNA | AK133534.1 | - | 1 | 2480 | 79 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 81 | Ccdc81 | 4921513D09Rik | -35.0kb | Sense | Chr. 7 - 89866148 - 89903629 on NCBI Build GRCm38 |
| Downstream | embryonic ectoderm development | Eed |
ENSMUSG00000039373, l(7)5Rn, l7Rn5, lusk |
13.5kb | Sense | Chr. 7 - 89954654 - 89981419 on NCBI Build GRCm38 |
Set Membership: |
|