Search
Search

Cyhr1| Gene Symbol: Cyhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276321.1 | NP_001263250.1 | 3 - 4 | 995 | 71 | |
| GenBank mRNA | AK080867.1 | - | 3 - 4 | 1016 | 71 | |
| AK129156.1 | - | 2 - 3 | 2102 | 71 | ||
| AK211953.1 | - | 1 - 2 | 69 | 71 | ||
| BC062184.1 | - | 3 - 4 | 984 | 6515695 | 71 | |
| BC082772.1 | - | 3 - 4 | 995 | 6508580 | 71 | |
Set Membership: |
|